Skip to Content
Merck

Skip To

474500

4-Methylumbelliferyl-N-acetyl-α-D-glucosaminide

Single spot by TLC, solid, N-acetyl-α-D-glucosaminidase substrate, Calbiochem®

Synonym(s):

4-Methylumbelliferyl-N-acetyl-α-D-glucosaminide, 4-Methylumbelliferyl-2-acetamido-2-deoxy-α-D-glucopyranoside

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg

Available to ship TODAYfromAreál Kühne+Nagel spol. s r.o.

CZK 4,540.00

About This Item

Empirical Formula (Hill Notation):
C18H21NO8
CAS Number:
Molecular Weight:
379.36
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.21
Form:
solid
Storage condition:
OK to freeze, desiccated, protect from light

CZK 4,540.00


Available to ship TODAYDetails


Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

4-Methylumbelliferyl-N-acetyl-α-D-glucosaminide, Ultrasensitive, fluorogenic substrate for assay of N-acetyl-α-D-glucosaminidase. Can be used to test for Sanfilippo syndrome B.

Quality Segment

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated, protect from light

impurities

<0.05% 4-methylumbelliferone, <0.2% β-anomer

color

white

solubility

DMF: soluble, DMSO: soluble

shipped in

ambient

storage temp.

10-30°C

SMILES string

N([C@H]1[C@H](O[C@@H]([C@H]([C@@H]1O)O)CO)Oc2cc3[o][c](cc(c3cc2)C)=O)C(=O)C

InChI

1S/C18H21NO8/c1-8-5-14(22)26-12-6-10(3-4-11(8)12)25-18-15(19-9(2)21)17(24)16(23)13(7-20)27-18/h3-6,13,15-18,20,23-24H,7H2,1-2H3,(H,19,21)/t13-,15-,16-,17-,18+/m1/s1

InChI key

QCTHLCFVVACBSA-FGTAOJJYSA-N

General description

Ultrasensitive, fluorogenic substrate for assay of N-acetyl-α-D-glucosaminidase. Can be used to test for Sanfilippo B syndrome. Excitation maximum: 360 nm; emission maximum: 440 nm.

Preparation Note

Unstable in solution; reconstitute just prior to use.

Analysis Note

single spot by TLC

Other Notes

Marsh, J., and Fensom, A.H. 1985. Clin. Genet.27, 258.
Chow, P., and Weissmann, B. 1981. Carbohydr. Res. 96, 87.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
474426474422474502
form

solid

form

solid

form

solid

form

crystalline solid

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

10-30°C

storage temp.

10-30°C

storage temp.

−20°C

storage temp.

−20°C

solubility

DMF: soluble, DMSO: soluble

solubility

methanol: soluble

solubility

DMF: 50 mg/mL

solubility

methanol: soluble, water: soluble

storage condition

OK to freeze, protect from light, desiccated

storage condition

OK to freeze, desiccated, protect from light

storage condition

OK to freeze, desiccated, protect from light

storage condition

OK to freeze, desiccated, protect from light


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters