Skip to Content
Merck
All Photos(1)

Documents

448104

Sigma-Aldrich

c-Met/RON Dual Kinase Inhibitor

The c-Met/RON Dual Kinase Inhibitor, also referenced under CAS 913376-84-8, controls the biological activity of c-Met/RON.

Synonym(s):

c-Met/RON Dual Kinase Inhibitor, Met Kinase Inhbitor IV, Ron Inhibitor I, N-(3-Fluoro-4-(7-methoxy-4-quinolinyl)phenyl)-1-(2-hydroxy-2-methylpropyl)-5-methyl-3-oxo-phenyl-2,3-dihydro-1H-pyrazole carboxamide, Compound I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H29FN4O5
CAS Number:
Molecular Weight:
556.58
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light tan

solubility

DMSO: 40 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable quinoline compound that acts as a potent inhibitor against the kinase activities of HGF (Cat. No. 375228) receptor c-Met and MSP (Macrophage Stimulating Protein) receptor RON (IC50 = 4 and 9 nM, respectively), while exhibiting much weaker potency toward Lck, Tie2, Src, BTK, IGFR, or VEGFR2 (IC50 = 160, 400, 410, 710, 1000, and 1900 nM, respectively), and little or no activity against B-Raf and more than 20 other kinases IC50 >1 µM). Shown to effectively supress (IC50<100 nM) the ligand-induced autophosphorylations of wild-type c-Met (in HT-29 and BxPC3 cultures) and RON (in NIH3T3 RON and BxPC3 cultures) as well as the ligand-independent autophosphorylations of the constitutively active mutants TPR-Met and RONΔ160 (in NIH3T3 TRP-Met and HT-29 cultures, respectively). Reported to inhibit NIH3T3 TRP-Met and U-87 tumor growth in mice in vivo in a dose-dependent manner (complete inhibition via daily p.o. dose of 100 mg/kg), but is less efficacious against HT-29, whose in vivo tumor growth is also dependent on V600E B-raf.
A cell-permeable quinoline compound that acts as a potent inhibitor against the kinase activities of HGF receptor c-Met and MSP (Macrophage Stimulating Protein) receptor RON (IC50 = 4 and 9 nM, respectively), while exhibiting much weaker potency toward Lck, Tie2, Src, BTK, IGFR, or VEGFR2 (IC50 = 160, 400, 410, 710, 1000, and 1900 nM, respectively), and little or no activity against B-Raf and more than 20 other kinases IC50 >1 M). Shown to effectively supress (IC50<100 nM) the ligand-induced autophosphorylations of wild-type c-Met (in HT-29 and BxPC3 cultures) and RON (in NIH3T3 RON and BxPC3 cultures) as well as the ligand-independent autophosphorylations of the constitutively active mutants TPR-Met and RONΔ160 (in NIH3T3 TRP-Met and HT-29 cultures, respectively). Reported to inhibit NIH3T3 TRP-Met and U-87 tumor growth in mice in vivo in a dose-dependent manner (complete inhibition via daily p.o. dose of 100 mg/kg), but is less efficacious against HT-29, whose in vivo tumor growth is also dependent on V600E B-raf.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Zhang, Y., et al. 2008. Cancer Res.68, 6680.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yihong Zhang et al.
Cancer research, 68(16), 6680-6687 (2008-08-15)
Recepteur d'origine nantais (RON) is a receptor tyrosine kinase closely related to c-Met. Both receptors are involved in cell proliferation, migration, and invasion, and there is evidence that both are deregulated in cancer. Receptor overexpression has been most frequently described
Kentaro Kajiwara et al.
Life science alliance, 4(4) (2021-02-13)
Compensatory growth of organs after loss of their mass and/or function is controlled by hepatocyte growth factor (HGF), but the underlying regulatory mechanisms remain elusive. Here, we show that CUB domain-containing protein 1 (CDCP1) promotes HGF-induced compensatory renal growth. Using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service