Skip to Content
Merck
All Photos(1)

Documents

437632

Sigma-Aldrich

Lipid A, Salmonella minnesota Re 595

Prepared by acid hydrolysis of LPS from Salmonella minnesota Re 595.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352211
NACRES:
NA.77

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

<0.2% 2-keto-3-deoxyoctonate

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

Prepared by acid hydrolysis of LPS from Salmonella minnesota Re 595. Component responsible for endotoxic properties of gram-negative bacterial lipopolysaccharides. Shows considerable similarity in structure and composition among enterobacteriaceae and many other bacterial families.
Prepared by acid hydrolysis of LPS from Salmonella minnesota Re 595. Component responsible for endotoxic properties of gram-negative bacterial lipopolysaccharides. Shows considerable similarity in structure and composition among enterobacteriaceaeand many other bacterial families.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Component responsible for endotoxic properties of gram-negative bacterial lipopolysaccharides.
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 1 month at 4°C.

Other Notes

Morrison, D.C., and Leive, L. 1975. J. Biol. Chem.250, 2911.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service