Skip to Content
Merck

Skip To

420141

JAK2 Inhibitor V, Z3

The JAK2 Inhibitor V, Z3, also referenced under CAS 195371-52-9, controls the biological activity of JAK2. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

JAK2 Inhibitor V, Z3, 2-Methyl-1-phenyl-4-pyridin-2-yl-2-(2-pyridin-2-ylethyl)butan-1-one

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C23H24N2O
CAS Number:
Molecular Weight:
344.45
UNSPSC Code:
12352200
Assay:
≥97% (HPLC)
Form:
oil
Storage condition:
OK to freeze, desiccated (hygroscopic), protect from light


Quality Segment

assay

≥97% (HPLC)

form

oil

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated (hygroscopic), protect from light

color

colorless

solubility

DMSO: 10 mg/mL, ethanol: 10 mg/mL

shipped in

wet ice

storage temp.

2-8°C

InChI

1S/C23H24N2O/c1-23(15-13-20-11-5-7-17-24-20,16-14-21-12-6-8-18-25-21)22(26)19-9-3-2-4-10-19/h2-12,17-18H,13-16H2,1H3

InChI key

SETYDCSRABYHSW-UHFFFAOYSA-N

General description

A cell-permeable pyridinylbutanone compound that inhibits cellular autophosphorylation of both the wild-type Jak2 and the constitutively active V167F mutant (IC50 = 15 & 28 µM, respectively, with WT- & V617F-transfected BSC-40 cells), while exhiting no effect against cellular Tyk2 autophosphorylation in COS cells or c-Src kinase in cell-free kinase assays. Z3 suppresses the Jak2-V167F-dependent proliferation of the erythroleukemia cell line HEL by blocking the G1-S transition and is shown to effectively reduce the cytokine-stimulated growth of both Jak2-V617F-positive and Jak2-F537I-positive primary hematopoietic progenitor cells from myeloproliferative disorder patients.

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Sayyah, J., et al. 2008. Mol. Cancer Ther.7, 2308.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters