Skip to Content
Merck
All Photos(3)

Documents

4104A

Millipore

Centrifree® Ultrafiltration Centrifugal Filters

new

non-IVD, MWCO 30KDa, sample volume 1mL, Ultracel® PL regenerated cellulose membrane

Synonym(s):

Centrifuge concentrator filter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104916
NACRES:
NB.24

material

EPDM rubber O-ring
EPDM rubber seal
Ultracel® PL regenerated cellulose membrane
polycarbonate base
polyethylene cup
styrene-acrylonitrile (SAN) reservoir

Quality Level

sterility

non-sterile

manufacturer/tradename

Centrifree®

device volume

1 mL

pore size

30 kDa pore size (MWCO)

shipped in

ambient

General description

The Centrifree® Ultrafiltration device with Ultracel regenerated cellulose membrane is ideal for separating free from bound microsolutes in serum, plasma, and other biological samples. The device is for research only, and holds a sample volume of 1 mL with a hold-up volume of 10 µL.

Application

  • Separate unbound (free) therapeutic drugs, testosterone, thyroxin or other bioactives
  • Binding Studies
  • New Drug Investigation
  • Facilitate deproteinization to reduce complexity or matrix interference of biological samples

Features and Benefits

  • Sample volumes of 0.15mL- 1mL, with 10uL hold up volume for high concentration factor
  • Ultracel® regenerated cellulose membrane, for high protein retention and accuracy

Legal Information

CENTRIFREE is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service