Skip to Content
Merck

Skip To

405210

ID-8

A cell-permeable indole compound that, when added to murine D3ES ESC (embryonic stem cell) cultures in the absence of serum and MEF (murine embryonic fibroblast) feeder cells, serves as a cheaper, chemical alternative to LIF

Synonym(s):

ID-8, 1-(4-Methoxyphenyl)-2-methyl-3-nitro-indol-6-ol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C16H14N2O4
CAS Number:
Molecular Weight:
298.29
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.51
Assay:
≥95% (HPLC)
Form:
powder
Storage condition:
OK to freeze, protect from light
Product 405210 is not currently sold in your country. Contact Technical Service
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

yellow

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[N+](=O)([O-])c1c2c([n](c1C)c3ccc(cc3)OC)cc(cc2)O

InChI

1S/C16H14N2O4/c1-10-16(18(20)21)14-8-5-12(19)9-15(14)17(10)11-3-6-13(22-2)7-4-11/h3-9,19H,1-2H3

InChI key

VVZNWYXIOADGSW-UHFFFAOYSA-N

General description

A cell-permeable indole compound that, when added (10 µM) to murine D3ES ESC (embryonic stem cell) cultures in the absence of serum and MEF (murine embryonic fibroblast) feeder cells, serves as a cheaper, chemical alternative to LIF (1,000 units/ml) for expanding ESCs in their undifferentiated state. Long-term murine ESCs maintained under ID-8 are capable of differentiating and EBs (Embryoid Bodies) formation upon ID-8 removal. Also claimed to maintain human ESCs in their pluripotent state in the absence of MEF.

Packaging

Packaged under inert gas

Other Notes

Miyabayashi, T., et al. 2008. Biosci. Biotechnol. Biochem.72, 1242.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Still not finding the right product?

Explore all of our products under ID-8


Storage Class

11 - Combustible Solids

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters