Skip to Content
Merck
All Photos(1)

Key Documents

401125

Sigma-Aldrich

Goat Anti-Human IgA, α-Chain Specific

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
CZK 5,030.00

CZK 5,030.00


Estimated to ship onApril 17, 2025Details


Request a Bulk Order

Select a Size

Change View
5 ML
CZK 5,030.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.77

CZK 5,030.00


Estimated to ship onApril 17, 2025Details


Request a Bulk Order

biological source

goat

Quality Level

antibody form

purified antibody

antibody product type

secondary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

ambient

storage temp.

−20°C

target post-translational modification

unmodified

General description

Purified goat polyclonal antibody. Recognizes the α-chain of human IgA.
This Goat Anti-Human IgA, α-Chain Specific is validated for use in Immunoelectrophoresis for the detection of Human IgA, α-Chain Specific.

Application

Immunoelectrophoresis (see comments)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In sterile-filtered (0.22 µm) 500 mM NaCl, 50 mM Tris-HCl, pH 7.5.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Monospecific for human immunoglobulin A, α-chain as determined by immunoelectrophoresis with undiluted antibody against 2X pooled human serum and undiluted pooled human plasma. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service