Skip to Content
Merck

Skip To

401114

Human IgG, Plasma

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mg

Estimated to ship onJune 30, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 3,500.00

About This Item

NACRES:
NA.47
UNSPSC Code:
51111800
Clone:
-
Species reactivity:
-
Application:
Citations:
-

CZK 3,500.00


Estimated to ship onJune 30, 2026Details


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

(Human plasma)

Quality Segment

form

lyophilized

storage condition

OK to freeze

isotype

IgG

storage temp.

−20°C

General description

Native IgG from human plasma.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.


Storage Class

13 - Non Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library