Skip to Content
Merck
All Photos(2)

Key Documents

382192

Sigma-Aldrich

HMTase Inhibitor IV, UNC0638

The HMTase Inhibitor IV, UNC0638 controls the biological activity of HMTase. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

HMTase Inhibitor IV, UNC0638, DNA Methyltransferase Inhibitor III, DNA MTase Inhibitor III, EHMT1/GLP Inhibitor II, EHMT2/G9a Inhibitor IV, 2-Cyclohexyl-N-(1-isopropylpiperidin-4-yl)-6-methoxy-7-(3-(pyrrolidin-1-yl)propoxy)quinazolin-4-amine

Sign Into View Organizational & Contract Pricing

Select a Size

2 MG
CZK 3,340.00

CZK 3,340.00


Estimated to ship onApril 17, 2025Details


Request a Bulk Order

Select a Size

Change View
2 MG
CZK 3,340.00

About This Item

Empirical Formula (Hill Notation):
C30H47N5O2
CAS Number:
Molecular Weight:
509.73
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

CZK 3,340.00


Estimated to ship onApril 17, 2025Details


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

CC(C)N(CC1)CCC1NC2=NC(C3CCCCC3)=NC4=C2C=C(C(OCCCN5CCCC5)=C4)OC

InChI

1S/C30H47N5O2/c1-22(2)35-17-12-24(13-18-35)31-30-25-20-27(36-3)28(37-19-9-16-34-14-7-8-15-34)21-26(25)32-29(33-30)23-10-5-4-6-11-23/h20-24H,4-19H2,1-3H3,(H,31,32,33)

InChI key

QOECJCJVIMVJGX-UHFFFAOYSA-N

General description

A cell-permeable quinazolinamine compound that acts as a potent and reversible inhibitor of G9a and GLP HMTases (histone methyltransferases) (IC50 = ≤ 15 and 19 nM for G9a and GLP, respectively) and displays ~15-fold greater selectivity over DNMT1 and minimally blocks the activities of JMJD2E (IC50 = 4.66 µM) and SETD7, SETD8, PRMT3 and SUV39H2 (IC50 >10 µM). Shown to lower H3K9Me2 levels in MDA-MB231 cells (IC50 = 81 nM; EC50 = 11.2 µM for cell toxicity) and 6-fold more potent than the HMTase Inhibitor, BIX-01294, (Cat. No. 382190). Also, affects the activities of adrenergic α1A, adrenergic α1B and muscarinic M2 by 90%, 69% and 64% and a panel of 26-receptors and ion-channels by ≤ 30% at 1 µM.




This probe is supplied in conjunction with the Structural Genomics Consortium (SGC). For further characterization details, please visit the UNC0638 probe summary on the SGC website.

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Other Notes

Vedadi, M., et al. 2011. Nat. Chem. Biol.7, 566.

Liu, F., et al. 2011. J. Med. Chem.54, 6139.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service