Skip to Content
Merck
All Photos(1)

Key Documents

344580

Sigma-Aldrich

FtsZ Polymer Stabilizer, PC190723

Synonym(s):

FtsZ Polymer Stabilizer, PC190723, 3-((6-chlorothiazolo[5,4-b]pyridin-2-yl)methoxy)-2,6-difluorobenzamide, FtsZ Polymer-stabilizing Agent, Filamenting Temperature-Sensitive Mutant Z Polymer Stabilizer

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
CZK 5,900.00

CZK 5,900.00


Estimated to ship onApril 11, 2025Details



Select a Size

Change View
5 MG
CZK 5,900.00

About This Item

Empirical Formula (Hill Notation):
C14H8ClF2N3O2S
Molecular Weight:
355.75
UNSPSC Code:
12352200
NACRES:
NA.77

CZK 5,900.00


Estimated to ship onApril 11, 2025Details


Assay

≥97% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

tan

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A benzamide ether derivative that exhibits potent bactericidal activity against Staphylococcus aureus and Bacillus subtilis (minimum inhibitory concentration = 0.5 to 1 μg/ml), but does not affect E. coli, yeast, or human hepatocytes. Shown to reduce GTPase activity and induce FtsZ (a prokaryotic homologue of tubulin) polymerization in S. aureus in a dose-dependent manner. Reported to directly inhibit the GTPase activity of FtsZ in a concentration dependent manner (IC50 = 55 ng/ml). Shown to bind between the C-ternimal domain and helix 7 in a region analogous to the Taxol binding site of tubulin. Increases the survival rate to 100% in mice subjected to a lethal dose of S. aureus.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
FtsZ
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Elsen, N. L., et al. 2012. J. Am. Chem. Soc.134, 12342.
Andreu, J. M., et al. 2010. J. Biol. Chem.285, 14239.
Haydon, D. J., et al. 2008. Science, 321, 1673.
Sorto, N. A., et al. 2010. J. Org. Chem.75, 7946.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service