Skip to Content
Merck

344037

cFMS Receptor Inhibitor II - CAS 959860-85-6 - Calbiochem

The cFMS Receptor Inhibitor II, also referenced under CAS 959860-85-6, controls the biological activity of cFMS. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

cFMS Receptor Inhibitor II - CAS 959860-85-6 - Calbiochem, 4-(3,4-Dimethylanilino)-7-(4-pyridyl)quinoline-3-carboxamide, CSF-1 Receptor Inhibitor II

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C23H20N4O
CAS Number:
Molecular Weight:
368.43
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

yellow

solubility

DMSO: 25 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable anilinoquinoline compound that acts as a potent, active site-targeting inhibitor of MCSF receptor/cFMS (IC50 = 80 nM). Although equally potent as cFMS Receptor Inhibitor III (Cat. No. 344038) in inhibiting the cFMS-dependent growth of M-NSF-60 cells (IC50 = 0.1 µM), this inhibitor is much more selective against cFMS-independent growth of NSO cells (IC50 >50 µM).
A cell-permeable anilinoquinoline compound that acts as a potent, active site-targeting inhibitor of MCSF receptor/cFMS (IC50 = 80 nM). Although equally potent as cFMS Receptor Inhibitor III (Cat. No. 344038) in inhibiting the cFMS-dependent growth of M-NSF-60 cells (IC50 = 0.1 µM), this inhibitor is much more selective against cFMS-independent growth of NSO cells (IC50 >50 µM).

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Smalley, T.L. Jr., et al. 2007. Bioorg. Med. Chem. Lett.17, 6257.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library