Skip to Content
Merck

Skip To

341691

Ficoll® 400 Reagent, Molecular Biology Grade

Useful for the separation of lymphocyte subpopulations.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 g

Available to ship TODAYfromAreál Kühne+Nagel spol. s r.o.

CZK 7,340.00
250 g

Estimated to ship onJune 08, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 31,300.00

About This Item

CAS Number:
UNSPSC Code:
12352201
NACRES:
NA.25
MDL number:

CZK 7,340.00


Available to ship TODAYDetails


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

Quality Segment

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: soluble

foreign activity

DNases, proteases, RNases, none detected

shipped in

ambient

storage temp.

15-25°C

General description

Useful for the separation of lymphocyte subpopulations.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Ficoll is a registered trademark of Cytiva

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F2637F5415F8016
grade

Molecular Biology

grade

Molecular Biology

grade

-

grade

-

solubility

water: soluble

solubility

-

solubility

water: soluble

solubility

-

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

200

form

solid

form

lyophilized powder

form

liquid

form

-

storage temp.

15-25°C

storage temp.

room temp

storage temp.

2-8°C

storage temp.

-

color

white

color

-

color

clear colorless

color

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters