Skip to Content
Merck

Skip To

324351

n-Dodecyl-β-D-glucopyranoside

Highly purified non-ionic detergent for solubilization of membrane-bound proteins in their native state.

Synonym(s):

n-Dodecyl-β-D-glucopyranoside, Dodecylglucoside

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 g

Estimated to ship onJune 08, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 7,890.00

About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.21
MDL number:

CZK 7,890.00


Estimated to ship onJune 08, 2026Details


Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥99% (HPLC)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated (hygroscopic)

color

white

CMC

130 μM

shipped in

ambient

storage temp.

2-8°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)OCCCCCCCCCCCC

InChI

1S/C18H36O6/c1-2-3-4-5-6-7-8-9-10-11-12-23-18-17(22)16(21)15(20)14(13-19)24-18/h14-22H,2-13H2,1H3/t14-,15-,16+,17-,18-/m1/s1

InChI key

PYIDGJJWBIBVIA-UYTYNIKBSA-N

General description

Highly purified non-ionic detergent for solubilization of membrane-bound proteins in their native state. CMC: 130 μM.

Preparation Note

Following reconstitution, refrigerate (4°C). Stock solutions are stable for up to 3 months at 4°C.
This detergent is not soluble above the CMC (130 µM or 0.45 mg/ml).

Other Notes

Deber, C.M., and Hsu, L.C. 1986. Biochem. Biophys. Res. Commun.134, 731.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Irritant (B)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
324374324355252718
assay

≥99% (HPLC)

assay

-

assay

≥98% (HPLC)

assay

≥99% (HPLC)

form

crystalline solid

form

solid

form

solid

form

lyophilized

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

CMC

130 μM

CMC

300 μM

CMC

100-600 μM

CMC

1.6 mM

color

white

color

white to off-white

color

white

color

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters