Skip to Content
Merck

Skip To

317201

DMBO

An oxazine that appears to mimic the pyranosidic moiety of heparin sulfate, and is shown to interact directly with several growth factors and cytokines including VEGF, TNF-α and heparin-binding-EGF in a surface plasmon resonance assay.

Synonym(s):

DMBO, CCDC-272761

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C22H25F2NO2
CAS Number:
Molecular Weight:
373.44
UNSPSC Code:
12352200
MDL number:
Assay:
≥95% (HPLC)
Form:
solid
Storage condition:
OK to freeze, protect from light


Quality Segment

assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

white

solubility

DMSO: soluble

shipped in

ambient

storage temp.

−20°C

SMILES string

COC1=CC=C(C2C3(CCCCC3)OC(C4=C(F)C=CC=C4F)NC2)C=C1

General description

An oxazine that appears to mimic the pyranosidic moiety of heparin sulfate, and is shown to interact directly with several growth factors and cytokines including VEGF, TNF-α and heparin-binding-EGF in a surface plasmon resonance assay. At concentrations of 10-80 µM, this compound dose-dependently inhibits the proliferation of VEGF-expressing LM8G7 cells (IC50 = 13 µM), and TNF-α-expressing OVSAHO cells (IC50 = 19 µM). At 195 µM, a 99% reduction in cell growth is observed in HB-EGF-expressing SKOV-3 cultures. This compound inhibits heparanase-mediated degradation of heparin sulfate in LM8G7 cells (IC50 = 65 µM and 104-206 µM, in an enzymatic and a cell based assay, respectively). At 0.5-5 µM, it markedly prevents the migration and invasion of the same culture, in a dose-dependent manner. In addition, it completely suppresses liver metastatic nodules of osteosarcoma mice injected with LM8G7 cells at 0.5 mg/kg without any signs of toxicity in vivo, and demonstrates a synergistic anti-tumor effect when dosed with heparin.

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Basappa, et al. 2010. Cancer Lett.279, 231.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters