Skip to Content
Merck
All Photos(1)

Documents

252926

Millipore

N-Acetylneuraminic Acid, 2,3-Dehydro-2-deoxy-, Sodium Salt

Inhibitor of bacterial, viral, and mammalian neuraminidases. Ki = 4 µM for influenza virus neuraminidase. Believed to act by binding to the catalytic site of neuraminidase as a transition state analog.

Synonym(s):

N-Acetylneuraminic Acid, 2,3-Dehydro-2-deoxy-, Sodium Salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16NNaO8
Molecular Weight:
313.24
UNSPSC Code:
12352201
NACRES:
NA.79

Quality Level

Assay

≥95% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

off-white to pale yellow

solubility

water: 1 mM

shipped in

ambient

storage temp.

−20°C

General description

A ganglioside neuraminidase inhibitor that is believed to act by binding to the catalytic site of neuraminidase. Reported to inhibit influenza virus neuraminidase (Ki = 4 µM). Also inhibits bacterial, viral, and mammalian neuraminidases.
Inhibitor of bacterial, viral, and mammalian neuraminidases. Ki = 4 µM for influenza virus neuraminidase. Believed to act by binding to the catalytic site of neuraminidase as a transition state analog.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Kopitz, J., et al. 1997. Eur. J. Cell. Biol.73, 1.
Holzer, C.T., et al. 1993. Glycoconj. J.10, 40.
Chong, A.K., et al. 1991. Biochem. Int.24, 165.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service