Skip to Content
Merck

Skip To

228111

Cholesterol, Highly Purified

≥90% (HPLC), ERRalpha ligand, powder

Synonym(s):

Cholesterol, Highly Purified

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 g

Estimated to ship TODAYfromAreál Kühne+Nagel spol. s r.o.

CZK 1,720.00

About This Item

Empirical Formula (Hill Notation):
C27H46O
CAS Number:
Molecular Weight:
386.65
UNSPSC Code:
12352211
NACRES:
NA.77
MDL number:
eCl@ss:
39023139
Assay:
≥90% (HPLC)
Form:
powder
Storage condition:
OK to freeze

CZK 1,720.00


Estimated to ship TODAYDetails


Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Cholesterol, Highly Purified, Suitable for use in liposome preparations.

description

Merck USA index - 14, 2201

Quality Segment

assay

≥90% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white to off-white

solubility

hot ethanol: 20 mg/mL, chloroform: 50 mg/mL, pyridine: 50 mg/mL

shipped in

ambient

storage temp.

10-30°C

SMILES string

O[C@H]1CC[C@@]2([C@@H]3[C@H]([C@H]4[C@@]([C@H](CC4)[C@@H](CCCC(C)C)C)(CC3)C)CC=C2C1)C

InChI

1S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9,18-19,21-25,28H,6-8,10-17H2,1-5H3/t19-,21+,22+,23-,24+,25+,26+,27-/m1/s1

InChI key

HVYWMOMLDIMFJA-DPAQBDIFSA-N

General description

Suitable for use in liposome preparations.

Biochem/physiol Actions

Reversible: no
Cell permeable: no
Primary Target
Suitable for use in liposome preparations
Product does not compete with ATP.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C3045PHR1533C2155000
form

powder

form

powder

form

powder

form

-

assay

≥90% (HPLC)

assay

≥99%

assay

-

assay

-

manufacturer/tradename

Calbiochem®

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

EDQM

Quality Level

100

Quality Level

200

Quality Level

300

Quality Level

-

storage temp.

10-30°C

storage temp.

−20°C

storage temp.

-10 to -25°C

storage temp.

2-8°C

storage condition

OK to freeze

storage condition

-

storage condition

-

storage condition

-


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters