Skip to Content
Merck
All Photos(1)

Key Documents

219334

Sigma-Aldrich

β-Catenin/Tcf Inhibitor V

BC21

Synonym(s):

β-Catenin/Tcf Inhibitor V, ICAT, Inhibitor of beta-catenin and Tcf

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
CZK 5,500.00

CZK 5,500.00


Estimated to ship onMarch 21, 2025Details


Request a Bulk Order

Select a Size

Change View
10 MG
CZK 5,500.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.28

CZK 5,500.00


Estimated to ship onMarch 21, 2025Details


Request a Bulk Order

Quality Level

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service