Skip to Content
Merck
All Photos(1)

Key Documents

217590

Sigma-Aldrich

Anti-CD40 Mouse mAb (EA-5)

liquid, ≥95% (SDS-PAGE), clone EA-5

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CZK 10,500.00

CZK 10,500.00


Estimated to ship onApril 28, 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
CZK 10,500.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CZK 10,500.00


Estimated to ship onApril 28, 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

EA-5, monoclonal

Assay

≥95% (SDS-PAGE)

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Anti-CD40, mouse monoclonal, clone EA-5, recognizes ~44 kDa CD40 in Raji cells. It is validated for use in Flow Cytometry, immunoprecipitation, & immunohistochemistry on frozen & paraffin sections.
Protein A purified mouse monoclonal antibody. Recognizes the ~44 kDa CD40 protein.
Recognizes the ~44 kDa CD40 protein in Raji cells.

Immunogen

transfected murine L cells expressing human CD40

Application

Flow cytometry (5 µg/ml)

Frozen Sections (see comments)

Immunoprecipitation (see comments)

Paraffin Sections (see comments)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 150 mM NaCl, 100 mM KCl, 50 mM sodium phosphate, 0.5 mg/ml gentamicin sulfate, pH 7.5.

Other Notes

FACS was performed on 5x105 cultured Raji human tumor cells. Optimal binding concentration tested was 5 µg/ml using FITC-conjugated goat anti-mouse IgG as a secondary antibody. This antibody has also been reported to work for frozen sections, immunoprecipitation, and paraffin sections. Variables associated with assay conditions will dictate the proper working dilution.
Fanslow, W.C., et al. 1992. J. Immunol.149, 655.
Law, C.L., et al. 1990. Leukemia4, 732.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service