Skip to Content
Merck
All Photos(1)

Documents

217274

Millipore

Carboxymethylcellulose, Sodium Salt, High Viscosity

Substrate for cellulase. High viscosity (2%, H2O, 25°C): 800 - 3100 mPas.

Synonym(s):

Carboxymethylcellulose, Sodium Salt, High Viscosity

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 5 mg/mL

shipped in

ambient

storage temp.

15-25°C

InChI

1S/C6H12O6.C2H4O2.Na/c7-1-3(9)5(11)6(12)4(10)2-8;1-2(3)4;/h1,3-6,8-12H,2H2;1H3,(H,3,4);

InChI key

DPXJVFZANSGRMM-UHFFFAOYSA-N

General description

Substrate for cellulase. Viscosity: (2%, H2O, 25°C): 800 - 3100 mPas.
Substrate for cellulase. Viscosity: (2%, H2O, 25°C): 800-3100 mPa s at 25°C, 2% H2O.

Warning

Toxicity: Standard Handling (A)

Other Notes

Christakopoulos, P., et al. 1999. Arch. Biochem. Biophys. 364, 61.
Blanco, A., et al. 1998. Appl. Microbiol. Biotechnol. 50, 48.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service