About This Item
Quality Segment
assay
>98% (SDS-PAGE)
form
lyophilized
specific activity
≥35,000 units/mg protein
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze, avoid repeated freeze/thaw cycles
solubility
water: 1 mg/mL
shipped in
wet ice
storage temp.
−20°C
General description
Physical form
Preparation Note
Other Notes
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| description Calmodulin, Porcine Brain, is a ubiquitous Ca2+-binding protein that serves as a physiological effector of a wide range of biological processes. Purified from porcine brain. | description Calmodulin is a ubiquitous Ca2+-binding protein that serves as a physiological effector of a wide range of biological processes. Purified from bovine brain. | description The Calmodulin Inhibitory Peptide controls the biological activity of Calmodulin. This small molecule/inhibitor is primarily used for Cell Signaling applications. | description Calmodulin, His•Tag Human, Recombinant, is a full-length, recombinant human calmodulin fused to a His•Tag sequence at the N-terminus. Contains four functional Ca2+-binding sites. |
| assay >98% (SDS-PAGE) | assay ≥95% (SDS-PAGE) | assay ≥95% (HPLC) | assay >90% (SDS-PAGE) |
| form lyophilized | form lyophilized | form lyophilized | form liquid |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −70°C |
| storage condition OK to freeze, avoid repeated freeze/thaw cycles | storage condition OK to freeze | storage condition OK to freeze, desiccated (hygroscopic) | storage condition avoid repeated freeze/thaw cycles, OK to freeze |
| specific activity ≥35,000 units/mg protein | specific activity ≥250,000 units/mg protein | specific activity - | specific activity - |
| solubility water: 1 mg/mL | solubility water: soluble, aqueous buffer: soluble | solubility water: 50 mg/mL | solubility - |
Storage Class
11 - Combustible Solids
WGK
WGK 1
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



