Skip to Content
Merck

208780

Calmodulin, Biotinylated, Porcine Brain

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

>98% (SDS-PAGE)

Quality Segment

form

lyophilized

specific activity

≥35,000 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

solubility

water: 1 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

Calmodulin (CaM) purified from porcine brain and biotinylated using NHS ester-activated biotin with an 8 atom spacer arm. A ubiquitous, calcium-binding protein that interacts with and regulates a wide variety of proteins, thus affecting different cellular functions and calcium-calmodulin-dependent signaling pathways. CaM mediates several biological processes, such as inflammation, apoptosis, muscle contraction, intracellular movement, short-term and long-term memory, nerve growth, and the immune response. Useful for enzyme activation, CaM binding studies, and general assessment of CaM-dependent signaling pathways. Biotin/Protein Ratio: 0.5-1.0 as determined by HABA assay.

Physical form

Lyophilized from 10 mM sodium phosphate buffer, 5% sucrose, pH 7.0.

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Billingsley, M.L., et al. 1985. Proc. Natl. Acad. Sci. USA82, 7585.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

10-13 - German Storage Class 10 to 13



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library