Skip to Content
Merck
All Photos(1)

Key Documents

203989

Sigma-Aldrich

S-Butyrylthiocholine Iodide

S-Butyrylthiocholine Iodide, CAS 1866-16-6, is a chromogenic substrate for cholinesterases. Its cleavage is measured colorimetrically at 405 nm.

Synonym(s):

S-Butyrylthiocholine Iodide

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
CZK 236,500.00

CZK 236,500.00


Estimated to ship onMarch 27, 2025Details


Request a Bulk Order

Select a Size

Change View
1 KG
CZK 236,500.00

About This Item

Empirical Formula (Hill Notation):
C9H20INOS
CAS Number:
Molecular Weight:
317.23
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.77

CZK 236,500.00


Estimated to ship onMarch 27, 2025Details


Request a Bulk Order

Quality Level

Assay

≥99% (silver nitrate titration)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

color

white

solubility

water: 1 mg/mL

absorption

≤0.20 at 405 nm (Free mercaptan, 1 cm)

shipped in

ambient

storage temp.

2-8°C

SMILES string

[I-].S(CC[N+](C)(C)C)C(=O)CCC

InChI

1S/C9H20NOS.HI/c1-5-6-9(11)12-8-7-10(2,3)4;/h5-8H2,1-4H3;1H/q+1;/p-1

InChI key

WEQAAFZDJROSBF-UHFFFAOYSA-M

General description

Chromogenic substrate for cholinesterases. Cleavage is measured colorimetrically at 405 nm.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Johnson, G., and Moore, S.W. 2000. Mol. Immunol.37, 707.
Hay, D.L., et al. 1983. Clin. Chem.29, 1065

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service