Skip to Content
Merck

Skip To

203189

Biotin-X-NHS, Water-Soluble

Synonym(s):

Biotin-X-NHS, Water-Soluble, Sulfobiotin-X-NHS, Sulfosuccinimidyl-6-(biotinamido)hexanoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
50 mg

Available to ship TODAYfromAreál Kühne+Nagel spol. s r.o.

CZK 19,200.00

About This Item

Empirical Formula (Hill Notation):
C20H29N4O9S2 · Na
Molecular Weight:
556.59
NACRES:
NA.41
UNSPSC Code:
41116133
Assay:
≥94% (HPLC)
Form:
solid

CZK 19,200.00


Available to ship TODAYDetails


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

(Synthetic chemical)

Quality Segment

assay

≥94% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated

color

off-white to tan

solubility

DMSO: 13 mg/mL, distilled water: 5.5 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

Water solubility is extremely convenient, eliminating the need to dissolve the reagent in an organic solvent such as dimethylformamide (DMF) and dimethylsulfoxide (DMSO) prior to use.
Readily couples biotin to primary amino groups of proteins under mild alkaline conditions. Ideally suited for labeling antibodies, hormones, or other polypeptide molecules containing lysine residues. Extended spacer arm provides maximum accessibility of biotin for avidin conjugates.

Preparation Note

Unstable in aqueous solution; reconstitute just prior to use. Following reconstitution in DMSO, Aliquot and freeze (-20°C). DMSO stock solutions are stable for up to 1 month -20°C.

Other Notes

Gretch, D.R., et al. 1987. Anal. Biochem.163, 270.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
203118QBD10200B1022
assay

≥94% (HPLC)

assay

-

assay

>90%

assay

≥90% (TLC)

solubility

DMSO: 13 mg/mL, distilled water: 5.5 mg/mL

solubility

DMSO: 10 mg/mL, distilled water: 4 mg/mL

solubility

-

solubility

H2O: 10 mg/mL, DMF: 50 mg/mL

Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

200

form

solid

form

solid

form

solid or viscous liquid

form

powder

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

biological source

(Synthetic chemical)

biological source

(Synthetic chemical)

biological source

-

biological source

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters