Skip to Content
Merck

Skip To

20-110

Src Manganese/ATP Cocktail

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 mL

Available to ship TODAYfromMerck (C/O Rhenus Logistics)

€161.00

About This Item

UNSPSC Code:
41116012
NACRES:
NA.41
eCl@ss:
32160405
Conjugate:
unconjugated
Technique(s):
activity assay: suitable (kinase)
Application:
activity assay
Citations:
-

€161.00


Available to ship TODAYDetails


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


conjugate

unconjugated

Quality Level

form

liquid

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SRC(6714)

Application

For use in kinase assays.
Research Category
All

Physical form

20mM MOPS, pH 7.2, 25mM β-glycerol phosphate, 5mM EGTA, 1mM sodium orthovanadate, 1mM dithiothreitol

Preparation Note

1 year at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
20-1132661671.12237
conjugate

unconjugated

conjugate

-

conjugate

-

conjugate

-

shipped in

wet ice

shipped in

wet ice

shipped in

-

shipped in

-

Gene Information

human ... SRC(6714)

Gene Information

-

Gene Information

-

Gene Information

-

technique(s)

activity assay: suitable (kinase)

technique(s)

activity assay: suitable (kinase)

technique(s)

-

technique(s)

-

UniProt accession no.

P12931

UniProt accession no.

-

UniProt accession no.

-

UniProt accession no.

-

form

liquid

form

liquid

form

chips

form

-


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
20-11004053252736452
Z561266-1EA04061833310489
BAB620090100-1EA04065269234838
Z423416-1EA04061833310045
Z556882-1EA04061832133225
Z423416-25EA04061833580578
SYNC151110-1EA04061832190303
SYNC171120-1EA04061832185835
SYNC571120-1EA04061832186436
Z640948-20EA04061831869248
Z744927-1EA04065272811392
Z744931-1EA04065272811477
Z744926-1EA04065272811378
Z744930-1EA04065272811453
Z676446-1EA04061833310939
WHA1034467204061831624007
WHA1034557204061831624021
WHA1034557604061831624038
GF90913855-1EA04061821162663
Z744932-1EA04065272811491
Z744928-1EA04065272811415
SYNP160020-1EA04065270932044
WHA1034467604061831624014
SYNF580020-1EA04065270931733
BAF378741100-20EA04065269239499