Skip to Content
Merck
All Photos(1)

Key Documents

197330

Sigma-Aldrich

Bcl-2 Inhibitor - CAS 383860-03-5 - Calbiochem

The Bcl-2 Inhibitor, also referenced under CAS 383860-03-5, controls the biological activity of Bcl-2. This small molecule/inhibitor is primarily used for Activators/Inducers applications.

Synonym(s):

Bcl-2 Inhibitor - CAS 383860-03-5 - Calbiochem, 2,9-Dimethoxy-11,12-dihydrodibenzo[c,g][1,2]-diazocine 5,6-dioxide (A) and 5,5ʹ-Dimethoxy-2,2ʹ-dinitrosobenzyl (B)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16N2O4
CAS Number:
Molecular Weight:
300.31
UNSPSC Code:
12352200

Assay

≥97% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

green

solubility

DMSO: 2.5 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

Contains a mixture of two tautomers (A and B). Potent, cell-permeable Bcl-2 inhibitor that competes with Bak BH3 peptide for binding to Bcl-2 and Bcl-XLin vitro (IC50 = 10 µM and 7 µM, respectively). Shown to reduce cell viability (IC50 = 10 µM) and growth in HL60 cells with high levels of Bcl-2 expression (IC50 = 4 µM). Induces apoptosis in a dose-dependent manner in cancer cells expressing high levels of Bcl-2, but has little or no effect on cells that express low levels of Bcl-2.
Contains mixture of two tautomers (A and B). A potent, cell-permeable, Bcl-2 inhibitor that competes with Bak BH3 peptide for binding to Bcl-2 and Bcl-xLin vitro (IC50 = 10 µM and 7 µM, respectively). Reduces cell viability and growth in the HL-60 cell line with high Bcl-2 expression (IC50 = 4 µM). Also induces apoptosis in a dose-dependent manner in cancer cells exhibiting high Bcl-2 expression with little effect on cancer cells with low Bcl-2 expression.

Biochem/physiol Actions

Cell permeable: yes
Reversible: no
Target IC50: 10 µM and 7 µM for competing with Bak BH3 peptide for binding to Bcl-2 and Bcl-xL in vitro, respectively; 4 µM for reducing cell viability and growth in the HL-60 cell line with high Bcl-2 expression

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution aliquot and freeze under inert gas (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Enyedy, I.J., et al. 2001. J. Med. Chem.44, 4313.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service