Skip to Content
Merck

Skip To

189732

Streptavidin, Alkaline Phosphatase Conjugate

This Streptavidin, Alkaline Phosphatase Conjugate is validated for use in ELISA, Immunoblotting, Immunohistochemistry.

Synonym(s):

Alkaline phosphatase-streptavidin, Streptavidin conjugate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
2 mL
Check Cart for Availability
CZK 10,300.00

About This Item

NACRES:
NA.47
UNSPSC Code:
41116133

CZK 10,300.00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

(Streptomyces avidinii)

Quality Segment

form

liquid

contains

≤0.1% sodium azide as preservative

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

dilution

(ELISA (1:1000-1:2500)
Immunoblotting (1:500-1:1250, colorimetric; 1:100-1:2500, chemiluminescence)
Immunohistochemistry (1:100-1:300))

shipped in

ambient

storage temp.

2-8°C

General description

Streptavidin conjugated to a highly purified alkaline phosphatase from calf intestine by a modified glutaraldehyde method. Specific for biotin, biotinylated proteins, or biotinylated proteins.

Application

ELISA (1:1000-1:2500)

Immunoblotting (1:500-1:1250, colorimetric; 1:100-1:2500, chemiluminescence)

Immunohistochemistry (1:100-1:300)

Packaging

Please refer to vial label for lot-specific concentration.

Physical form

In proprietary buffer.

Other Notes

Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1108915300111089161001189733
Quality Level

100

Quality Level

-

Quality Level

-

Quality Level

100

form

liquid

form

liquid

form

solution

form

liquid

biological source

(Streptomyces avidinii)

biological source

-

biological source

-

biological source

(Streptomyces avidinii)

storage temp.

2-8°C

storage temp.

-10 to -25°C

storage temp.

2-8°C

storage temp.

2-8°C

shipped in

ambient

shipped in

-

shipped in

-

shipped in

wet ice

storage condition

do not freeze

storage condition

-

storage condition

-

storage condition

do not freeze




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters