Skip to Content
Merck
All Photos(2)

Key Documents

189512

Sigma-Aldrich

Autotaxin Inhibitor III, PF-8380

The Autotaxin Inhibitor III, PF-8380, also referenced under CAS 1144035-53-9, controls the biological activity of Autotaxin. This small molecule/inhibitor is primarily used for Membrane applications.

Synonym(s):

Autotaxin Inhibitor III, PF-8380, 6-(3-(Piperazin-1-yl)propanoyl)-benzo[d]oxazol-2(3H)-one, Atx Inhibitor III, PF-8380

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H21Cl2N3O5
CAS Number:
Molecular Weight:
478.33
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.28
Product 189512 is not currently sold in your country. Contact Technical Service

Quality Level

Assay

≥95% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

tan

solubility

DMSO: 100 mg/mL

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=C(OCC1=CC(Cl)=CC(Cl)=C1)N2CCN(CCC(C3=CC(OC(N4)=O)=C4C=C3)=O)CC2

InChI

1S/C22H21Cl2N3O5/c23-16-9-14(10-17(24)12-16)13-31-22(30)27-7-5-26(6-8-27)4-3-19(28)15-1-2-18-20(11-15)32-21(29)25-18/h1-2,9-12H,3-8,13H2,(H,25,29)

InChI key

JMSUDQYHPSNBSN-UHFFFAOYSA-N

General description

An orally bioavailable piperazinylbenzoxazolone compound that acts as a substrate competitive and tight-binding inhibitor of autotaxin activity {IC50 = 2.8 and 1.7 nM for recombinant human enzyme-β isoform employing FS-3 and LPC (lysophosphatidylcholine) as substrates, respectively; 1.16 and 1.15 nM for rat/murine enzyme-FS-3 and fetal fibroblast cell-LPC; 101 nM for human whole blood}. Displays desirable pharmacokinetics properties and efficiently blocks inflammation-induced LPA (lysophosphatidic acid) production both in plasma and at the site of inflammation by 95% in rat adjuvant-induced arthritis model (30 mg/kg, p.o.).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Gierse, J., et al. 2010. J. Pharmacol. Exp. Ther.334, 310.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service