Skip to Content
Merck
All Photos(1)

Key Documents

189510

Sigma-Aldrich

Autotaxin Inhibitor I, S32826

The Autotaxin Inhibitor I, S32826, also referenced under CAS 1103672-43-0, controls the biological activity of Autotaxin.

Synonym(s):

Autotaxin Inhibitor I, S32826, ATX Inhibitor I, (4-(Tetradecanoylamino)benzyl)phosphonic acid disodium, dihydrate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
CZK 4,700.00

CZK 4,700.00


Estimated to ship onMarch 21, 2025Details



Select a Size

Change View
5 MG
CZK 4,700.00

About This Item

Empirical Formula (Hill Notation):
C21H34NNa2O4P · 2H2O
CAS Number:
Molecular Weight:
477.48
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

CZK 4,700.00


Estimated to ship onMarch 21, 2025Details


Quality Level

Assay

≥97% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

water: 0.5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[P](=O)([O-])([O-])Cc1ccc(cc1)NC(=O)CCCCCCCCCCCCC

InChI

1S/C21H36NO4P.2Na/c1-2-3-4-5-6-7-8-9-10-11-12-13-21(23)22-20-16-14-19(15-17-20)18-27(24,25)26;;/h14-17H,2-13,18H2,1H3,(H,22,23)(H2,24,25,26);;/q;2*+1/p-2

InChI key

DGRFALMFDGBLCP-UHFFFAOYSA-L

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service