Skip to Content
Merck

178196

α1-Antichymotrypsin, Human Plasma

Acute-phase protein present at ~45 mg/100 ml in plasma.

Synonym(s):

α1-Antichymotrypsin, Human Plasma, α1-Achy, α1-1X, α1-1X-Glycoprotein

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.77
MDL number:
Assay:
≥95% (SDS-PAGE)
Form:
solid
Storage condition:
OK to freeze
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (SDS-PAGE)

form

solid

specific activity

≥85%, pH 7.4, 25 °C (using Suc-Ala-Ala-Pro-Phe-pNA as substrate, inhibition of cathepsin G)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

aqueous buffer: soluble

shipped in

ambient

storage temp.

−20°C

General description

Acute-phase protein present at 45 mg/100 ml in plasma. Specific inhibitor of chymotrypsin-like serine proteinases.
Acute-phase protein present at ~45 mg/100 ml in plasma. Specific inhibitor of chymotrypsin-like serine proteinases. Molecular weight ranges from 65,000-68,000 daltons.
M.W. 65,000-68,000.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Chymotrypsin-like serine proteases
Product does not compete with ATP.
Reversible: no

Physical form

Lyophilized from 150 mM NaCl, 20 mM Tris-HCl, pH 7.4.

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for at up to 6 months at -20°C.
Please refer to vial label for lot-specific volume prior to lyophilization.
Prepared from plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Other Notes

Desrochers, P.E., et al. 1992. J. Biol. Chem.267, 5005.
Rayford, A., et al. 1992. J.Neurochem.58, 88.
Travis, J., et al. 1978. Biochemistry17, 5651.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Toxicity: Standard Handling (A)


Storage Class

13 - Non Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library