Skip to Content
Merck
All Photos(1)

Key Documents

17-671

Sigma-Aldrich

ChIPAb+ Trimethyl-Histone H4 (Lys20) - ChIP Validated Antibody and Primer Set, rabbit monoclonal

culture supernatant, from rabbit

Synonym(s):

H4K20me3, Histone H4 (tri methyl K20), H4 histone family, member A, histone 1, H4a, histone cluster 1, H4a

Sign Into View Organizational & Contract Pricing

Select a Size

10 ASSAYS
CZK 13,800.00

CZK 13,800.00


Estimated to ship onMarch 06, 2025Details


Request a Bulk Order

Select a Size

Change View
10 ASSAYS
CZK 13,800.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.52
clone:
monoclonal
application:
ChIP
IP
WB
species reactivity:
human, vertebrates, plant
technique(s):
ChIP: suitable
immunoprecipitation (IP): suitable
western blot: suitable
citations:
1

CZK 13,800.00


Estimated to ship onMarch 06, 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

culture supernatant

clone

monoclonal

species reactivity

human, vertebrates, plant

manufacturer/tradename

ChIPAb+
Upstate®

technique(s)

ChIP: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Related Categories

General description

All ChIPAb+ antibodies are individually validated for chromatin precipitation, every lot, every time. Each ChIPAb+ antibody set includes control primers (tested every lot by qPCR) to biologically validate your IP results in a locus-specific context. The qPCR protocol and primer sequences are provided, allowing researchers to validate ChIP protocols when using our antibody in their chromatin context. Each set also includes a negative control antibody to ensure specificity of the ChIP reaction.
The ChIPAb+ Trimethyl-Histone H4 (Lys20) set includes the Anti-trimethyl-Histone H4 (Lys20) antibody, a negative control supernatant, and qPCR primers which amplify a 110 bp region within the promoter of the humanβ-globin gene. The trimethyl histone H4 (Lys20) and negative control antibodies are supplied in a scalable "per ChIP" reaction size and can be used to functionally validate the precipitation of trimethyl-histone H4 (Lys20) associated chromatin.

Specificity

Not tested in other species. The immunogen sequence is identical in a wide range of animal and plant species, so broad cross-reactivity is expected.
Recognizes histone H4, Mr 11 kDa

Immunogen

Epitope: a.a. 18-22
The trimethyl histone H4 (Lys20) antibody is made against a synthetic peptide corresponding to amino acids 18-22 of histone H4. trimethylated on lysine 20.

Application

Chromatin Immunoprecipitation:
Sonicated chromatin prepared from HeLa cells (2 X 106 cell equivalents per IP) was subjected to chromatin immunoprecipitation using 10 μL of either a negative control supernatant or Anti-Trimethyl-Histone H4 (Lys20) antibody and the Magna ChIP A Kit (Part No. 17-610). Successful immunoprecipitation of trimethylhistone H4 (Lys20) associated DNA fragments was verified by qPCR using β-globin ChIP Primers versus Control Primers corresponding to the GAPDH promoter (Please see figures). Data is presented as percent input of each IP sample relative to input chromatin, with immunoprecipitated DNA from negative control supernatant shown as (-) and trimethyl histone H4 (Lys20) shown as (+).
Please refer to the EZ-Magna ChIP A (Cat. # 17-408) or EZ-ChIP (Cat. # 17-371) protocol for experimental details.

Western Blot Analysis:
Acid extracts from HeLa cells untreated or pre-absorbed with 1 μM of trimethylated (Lys20) Histone H4 peptide (Lanes 1 and 2 respectively) were probed with anti-trimethyl-Histone H4 (Lys20) (1:1,000) (Please see figures).

Dot Blot Analysis: Absurance Histone H3 Antibody Specificity Array (Cat. No. 16-667) and Absurance Histone H2A, H2B, H4 Antibody Specificity Array (Cat. No. 16-665), which contain histone peptides with various modifications were probed with Cat. No 04-079, Anti-trimethyl-Histone H4 (Lys20) ; dilution 1:500. Proteins were visualized using a Donkey anti-rabbit IgG conjugated to HRP and a chemiluminescence detection system.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Histones
This ChIPAb+ Trimethyl-Histone H4 (Lys20) -ChIP Validated Antibody & Primer Set conveniently includes the antibody & the specific control PCR primers.

Packaging

10 assays per set. Recommended use: ~10 μL antibody per chromatin immunoprecipitation (dependent upon biological context).

Quality

Chromatin Immunoprecipitation:
Sonicated chromatin prepared from HeLa cells (1 X 106 cell equivalents per IP) were subjected to chromatin immunoprecipitation using 10 µL of either a negative control supernatant, or Anti-Trimethyl-Histone H4 (Lys20) antibody and the Magna ChIP® A Kit (Cat. # 17-610).
Successful immunoprecipitation of trimethyl-histone H4 (Lys20)-associated DNA fragments was verified by qPCR using ChIP Primers B-Globin.
Please refer to the EZ-Magna ChIP A (Cat. # 17-408) or EZ-ChIP (Cat. # 17-371) protocol for experimental details.

Target description

~11 kDa

Physical form

Anti-trimethyl-Histone H4 (Lys20) (rabbit monoclonal IgG). One vial containing 100 μL of cell culture supernatant in 0.1% sodium azide. Store at -20°C.

Negative Control Supernatant. One vial containing 100 μL cell rabbit monoclonal culture supernatant containing 0.1% sodium azide. Store at -20°C.

ChIP Primers B-Globin. One vial containing 75 μL of 5 μM of each primer specific for human β-globin. Store at -20°C.
FOR: AGG ACA GGT ACG GCT GTC ATC
REV: TTT ATG CCC AGC CCT GGC TC

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Includes negative control rabbit antibody and control primers specific for human β-globin promoter.

Legal Information

MAGNA CHIP is a registered trademark of Merck KGaA, Darmstadt, Germany
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rita Rebollo et al.
PLoS genetics, 7(9), e1002301-e1002301 (2011-10-08)
The "arms race" relationship between transposable elements (TEs) and their host has promoted a series of epigenetic silencing mechanisms directed against TEs. Retrotransposons, a class of TEs, are often located in repressed regions and are thought to induce heterochromatin formation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service