Skip to Content
Merck
All Photos(1)

Documents

1.72577

SAFC

Sodium hydrogen carbonate

for table water, FCC, E 500

Synonym(s):

Sodium bicarbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaHCO3
CAS Number:
Molecular Weight:
84.01
Beilstein:
4153970
MDL number:
UNSPSC Code:
12352302
E Number:
E500
EC Index Number:
205-633-8

reg. compliance

FCC

Quality Level

form

powder

pH

8.6 (20 °C, 50 g/L in H2O)

pKa (25 °C)

(1) 6.37, (2) 10.25 (carbonic acid)

mp

270 °C (decomposition)

solubility

96 g/L

density

2.22 g/cm3 at 20 °C
2.16 g/mL at 25 °C (lit.)

application(s)

pharma/biopharma processes

storage temp.

2-30°C

SMILES string

[Na+].OC([O-])=O

InChI

1S/CH2O3.Na/c2-1(3)4;/h(H2,2,3,4);/q;+1/p-1

InChI key

UIIMBOGNXHQVGW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

We offer a range of high quality critical raw materials plus the documentation and expertise to help our customers meet stringent regulatory requirements. Our SAFC® portfolio of high-quality products withstands strict quality control procedures.

Application

Sodium hydrogen carbonate with a low bromide level can be used to remineralize desalinated sea water to produce table water.

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service