Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41106231
material
polystyrene plate
Quality Segment
agency
EP, USP
sterility
sterile; γ-irradiated
form
plate
feature
2D bar code, ready-to-use
packaging
pkg of (20/200 Plates per box)
D
55 mm
pH
(7.3 (in H2O))
application(s)
microbiology
pharmaceutical
surface monitoring
storage temp.
15-25°C
suitability
bacteria, fungi, molds, yeasts
General description
Tryptic Soy Contact Agar + LT - ICR is designed for the determination of the total microbial aerobic count on dry, sanitized surfaces and personnel in Clean rooms and Isolators. It is a complex medium for cultivation and isolation of a wide range of bacteria, yeasts and molds. The formulation of the Soybean Casein-Digest medium contains a combination of casein and soy peptones supplying organic nitrogen, particularly amino acids and longer-chained peptides. The medium is supplemented with Lecithin, polysorbate 80 (Tween®) and pyruvate to inactivate potential residues of disinfectants on dry surfaces. .
Application
Tryptic Soy Agar (TSA, Soybean Casein Digest Agar) is a complex medium for cultivation and isolation of a wide range of bacteria, yeasts and molds. The media finds its application in environmental monitoring by determining the total microbial aerobic count on dry, sanitized surfaces and personnel in Isolators and Clean Rooms
Packaging
1665020020-CN: 20 Plates per box
1665020200-CN: 200 Plates per box
1665020200-CN: 200 Plates per box
Legal Information
TWEEN is a registered trademark of Croda International PLC
Still not finding the right product?
Explore all of our products under Tryptic Soy Agar
Storage Class
13 - Non Combustible Solids
wgk
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.