Skip to Content
Merck
All Photos(1)

Key Documents

164640

Sigma-Aldrich

Aminopurvalanol A

A cell-permeable 2,6,9-trisubstituted purine analog that displays anti-mitotic as well as anti-tumor properties.

Synonym(s):

Aminopurvalanol A, (2R)-2-((6-((3-Amino-5-chlorophenyl)amino)-9-(1-methylethyl)-9H-purin-2-yl)amino)-3-methyl-1-butanol, NG-97

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H26ClN7O
CAS Number:
Molecular Weight:
403.91
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

DMSO: 40 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C19H26ClN7O/c1-10(2)15(8-28)24-19-25-17(23-14-6-12(20)5-13(21)7-14)16-18(26-19)27(9-22-16)11(3)4/h5-7,9-11,15,28H,8,21H2,1-4H3,(H2,23,24,25,26)/t15-/m0/s1

InChI key

RAMROQQYRRQPDL-HNNXBMFYSA-N

General description

A cell-permeable 2,6,9-trisubstituted purine analog that displays anti-mitotic as well as anti-tumor properties (GI50 = ~ 1.8 µM in the NCI 60-cell panel in vitro activity screen; potently inhibits the growth of KM12 colon cancer cells with a GI50 of 30 nM). Acts as a reversible and ATP-competitive inhibitor of Cdks (IC50 = 33 nM for Cdk1/cyclin B and Cdk2/cyclin A, 28 nM for Cdk2/cyclin E, and 20 nM for Cdk5/p35), and displays ~ 100-fold greater selectivity over a panel of kinases tested (IC50 ≥ 2.4 µM). Shown to induce cell differentiation by preferentially targeting the G2/M-phase and act intracellularly by inhibiting both Cdks and MAPKs.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Cdk1/cyclin B, Cdk2/cyclin A, Cdk2/cyclin E, and Cdk5/p35
Product competes with ATP.
Reversible: yes
Target IC50: 33 nM for Cdk1/cyclin B and Cdk2/cyclin A, 28 nM for Cdk2/cyclin E, and 20 nM for Cdk5/p35

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Breton, M.L., et al. 2003. Biochem. Biophys. Res. Commun.306, 880.
Knockaert, M., et al. 2002.Oncogene21, 6413.
Adachi, S., et al. 2001. Mol. Cell. Biol.21, 4929.
Knockaert, M., et al. 2000. Chem. Biol.7, 411.
Rosiana, G.R., et al. 1999. Proc. Natl. Acad. Sci. USA96, 4797.
Chang, Y.T., et al. 1999. Chem. Biol.6, 361.

Legal Information

Sold under license of U.S. Patent 6,255,485 and 6,617,331.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service