Skip to Content
Merck

Skip To

14-934-M

GCN2, active

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 μg

Estimated to ship onJune 03, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 13,600.00
250 μg

Estimated to ship onJune 05, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 86,800.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32
eCl@ss:
32160405
Biological source:
human

CZK 13,600.00


Estimated to ship onJune 03, 2026Details


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

mol wt

Mw 215 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

Gene Information

human ... EIF2AK4(440275)

General description

GCN2 (EIF2AK4) belongs to a family of kinases that phosphorylate the alpha subunit of eukaryotic translation initiation factor-2 (EIF2S1) to downregulate protein synthesis in response to varied cellular stresses.
Product Source: N-terminal GST-tagged, recombinant, human GCN2 full length, expressed by baculovirus in Sf21 insect cells. The recombinant protein contains the amino acid substitution E556G with reference to GenPept NP_001013725. E556G is reported in GenPept CAH10626.

Biochem/physiol Actions

Target Sub-Family: STE

Packaging

Also available in 250μg size as catalog # 14-934M and in bulk as catalog # 14-934-K.

Physical form

Protein in 50mM Tris/HCl pH7.5, 300mM NaCl, 0.1mM EGTA, 0.03% Brij-35, 270mM sucrose, 1mM benzamidine, 0.2mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Purified using glutathione sepharose.

Preparation Note

6 months at -70°C

Analysis Note

Routinely evaluated by phosphorylation of peptide RSRSRSRSRSRSRSR

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SRP5218MABN450HPA026776
technique(s)

activity assay: suitable (kinase)

technique(s)

-

technique(s)

western blot: suitable

technique(s)

immunofluorescence: 0.25-2 μg/mL, immunohistochemistry: 1:50-1:200

biological source

human

biological source

-

biological source

mouse

biological source

rabbit

Gene Information

human ... EIF2AK4(440275)

Gene Information

human ... EIF2AK4(440275)

Gene Information

human ... GCM2(9247)

Gene Information

human ... GCNT2(2651)

UniProt accession no.

Q9P2K8

UniProt accession no.

-

UniProt accession no.

O75603

UniProt accession no.

Q8N0V5

mol wt

Mw 215 kDa

mol wt

~132 kDa

mol wt

-

mol wt

-

NCBI accession no.

NP_001013725

NCBI accession no.

NM_001013703

NCBI accession no.

NP_004743

NCBI accession no.

-


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters