Skip to Content
Merck

Skip To

14-927-M

PAK1 Protein, active, 10 µg

Active, N-terminal FLAG-tagged, recombinant, human PAK1, amino acids 150-end. For use in Kinase Assays.

PAK1 Protein, active, 10 µg
1 of 1 reviewers received a sample product or took part in a promotion
Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 μg

Estimated to ship onMay 29, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 11,500.00
250 μg

Estimated to ship onJune 05, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 92,000.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.41
eCl@ss:
32160405
Biological source:
human
Recombinant:
expressed in baculovirus infected Sf21 cells
Mol wt:
Mw 46 kDa

CZK 11,500.00


Estimated to ship onMay 29, 2026Details


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

recombinant

expressed in baculovirus infected Sf21 cells

mol wt

Mw 46 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

General description

Product Source: N-terminal FLAG-tagged, recombinant, human PAK1 amino acids 150-end, expressed by baculovirus in Sf21 insect cells.
This protein is a family member of serine/threonine p21-activating kinases, known as PAK proteins. These proteins are critical effectors that link RhoGTPases to cytoskeleton reorganization and nuclear signaling, and they serve as targets for the small GTP binding proteins Cdc42 and Rac. This specific family member regulates cell motility and morphology. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.

Biochem/physiol Actions

Protein Target: PAK1

Packaging

Also available in 10μg size as catalog # 14-927 and in 250 μg as catalog # 14-927M.

Analysis Note

Routinely evaluated by phosphorylation of peptide RRRLSFAEPG.

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
14-452-M14-918-M14-777-M
recombinant

expressed in baculovirus infected Sf21 cells

recombinant

expressed in baculovirus infected Sf21 cells

recombinant

expressed in baculovirus infected Sf21 cells

recombinant

expressed in baculovirus infected Sf21 cells

biological source

human

biological source

human

biological source

human

biological source

human

technique(s)

activity assay: suitable (kinase)

technique(s)

activity assay: suitable (kinase)

technique(s)

activity assay: suitable (kinase)

technique(s)

activity assay: suitable (kinase)

mol wt

Mw 46 kDa

mol wt

Mw 59 kDa

mol wt

Mw 61 kDa

mol wt

Mw 69.3 kDa

UniProt accession no.

Q13153

UniProt accession no.

O15530

UniProt accession no.

P23458

UniProt accession no.

P53350, Q07832 (mouse), Q62673 (rat)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

Active Filters

  1. Toronto, ON
    • Reviews 4
    • Vote 1
    5 out of 5 stars.

    Active, good for kinase assays.

    Active, a little on the expensive side. Worked well for invitro kinase assays.

    Helpful?