Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
eCl@ss:
32160405
UNSPSC Code:
12352202
Recombinant:
expressed in baculovirus infected Sf21 cells
Pricing and availability is not currently available.
General description
50.0 kDa
FUNCTION: SwissProt: P08581 # Receptor for hepatocyte growth factor and scatter factor. Has a tyrosine-protein kinase activity.
SIZE: 1390 amino acids; 155527 Da
SUBUNIT: Heterodimer formed of an alpha chain (50 kDa) and a beta chain (145 kDa) which are disulfide linked. Binds PLXNB1 and GRB2. Interacts with SPSB1, SPSB2 and SPSB4 (By similarity). Interacts with INPP5D/SHIP1. When phosphorylated at Tyr-1356, interacts with INPPL1/SHIP2. Interacts with RANBP9 and RANBP10.
SUBCELLULAR LOCATION: Membrane; Single-pass type I membrane protein.
DISEASE: SwissProt: P08581 # Activation of MET after rearrangement with the TPR gene produces an oncogenic protein. & Defects in MET may be associated with gastric cancer. & Defects in MET are a cause of hepatocellular carcinoma (HCC) [MIM:114550]. & Defects in MET are a cause of hereditary papillary renal carcinoma (HPRC) [MIM:605074]; also known as papillary renal cell carcinoma 2 (RCCP2). HPRC is a form of inherited kidney cancer characterized by a predisposition to develop multiple, bilateral papillary renal tumors. The pattern of inheritance is consistent with autosomal dominant transmission with reduced penetrance. & Genetic variations in MET may be associated with susceptibility to autism type 1B (AUTS1B) [MIM:608636]. Autism is a neurodevelopmental disorder characterized by disturbance in language, perception and socialization. The disorder is classically defined by a triad of limited or absent verbal communication, a lack of reciprocal social interaction or responsiveness, and restricted, stereotypical, and ritualized patterns of interests and behavior.
SIMILARITY: SwissProt: P08581 ## Belongs to the protein kinase superfamily. Tyr protein kinase family. & Contains 3 IPT/TIG domains. & Contains 1 protein kinase domain. & Contains 1 Sema domain.
SIZE: 1390 amino acids; 155527 Da
SUBUNIT: Heterodimer formed of an alpha chain (50 kDa) and a beta chain (145 kDa) which are disulfide linked. Binds PLXNB1 and GRB2. Interacts with SPSB1, SPSB2 and SPSB4 (By similarity). Interacts with INPP5D/SHIP1. When phosphorylated at Tyr-1356, interacts with INPPL1/SHIP2. Interacts with RANBP9 and RANBP10.
SUBCELLULAR LOCATION: Membrane; Single-pass type I membrane protein.
DISEASE: SwissProt: P08581 # Activation of MET after rearrangement with the TPR gene produces an oncogenic protein. & Defects in MET may be associated with gastric cancer. & Defects in MET are a cause of hepatocellular carcinoma (HCC) [MIM:114550]. & Defects in MET are a cause of hereditary papillary renal carcinoma (HPRC) [MIM:605074]; also known as papillary renal cell carcinoma 2 (RCCP2). HPRC is a form of inherited kidney cancer characterized by a predisposition to develop multiple, bilateral papillary renal tumors. The pattern of inheritance is consistent with autosomal dominant transmission with reduced penetrance. & Genetic variations in MET may be associated with susceptibility to autism type 1B (AUTS1B) [MIM:608636]. Autism is a neurodevelopmental disorder characterized by disturbance in language, perception and socialization. The disorder is classically defined by a triad of limited or absent verbal communication, a lack of reciprocal social interaction or responsiveness, and restricted, stereotypical, and ritualized patterns of interests and behavior.
SIMILARITY: SwissProt: P08581 ## Belongs to the protein kinase superfamily. Tyr protein kinase family. & Contains 3 IPT/TIG domains. & Contains 1 protein kinase domain. & Contains 1 Sema domain.
Packaging
Available in 250 μg --call for pricing and availability and reference catalog number 14-816M when ordering the 250 μg size.
Preparation Note
6 months at -70°C
Analysis Note
For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
signalword
Warning
hcodes
Hazard Classifications
Skin Sens. 1
Storage Class
12 - Non Combustible Liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
