Skip to Content
Merck
All Photos(1)

Key Documents

14-782-M

Sigma-Aldrich

PI 3-Kinase (p110α(E542K)/p85α), Human, 10 µg

Complex of N-terminal 6His-tagged, recombinant, full-length human p110α containing the mutation E542K & untagged, recombinant, full-length human p85α. For use in Kinase Assays.

Synonym(s):

Active Kinase Protein, Human PI 3-Kinase

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
CZK 11,100.00
250 μG
CZK 88,100.00

CZK 11,100.00


Estimated to ship onMarch 19, 2025Details


Request a Bulk Order

Select a Size

Change View
10 μG
CZK 11,100.00
250 μG
CZK 88,100.00

About This Item

UNSPSC Code:
12352200
eCl@ss:
32160405
NACRES:
NA.32

CZK 11,100.00


Estimated to ship onMarch 19, 2025Details


Request a Bulk Order

biological source

human

Quality Level

100
300

recombinant

expressed in baculovirus infected Sf21 cells

mol wt

Mw 125.3 kDa (p110α; E542K)
Mw 83.7 kDa (p85α)

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

Gene Information

human ... PIK3CA(5290)

General description

Complex of N-terminal 6His-tagged, recombinant, full-length human p110α containing the mutation E542K and untagged, recombinant, full-length human p85α.
The E542K substitution is a somatic mutation in p110α that has been associated with tumours of the colon and brain. Combined in vitro and in vivo studies have shown that this mutation confers higher lipid kinase activity than wild type, and is able to induce oncogenic transformation.

Application

Complex of N-terminal 6His-tagged, recombinant, full-length human p110α containing the mutation E542K & untagged, recombinant, full-length human p85α. For use in Kinase Assays.

Packaging

Also available in 250 μg size --call for pricing and availability and reference catalog number 14-782M when ordering the 250 μg size.

Storage and Stability

Store at -70°C from date of shipment. For maximum recovery of product, centrifuge original vial prior to removing the cap.

Rapidly thaw the vial under cold water and immediately place on ice. Aliquot unused material into pre-chilled microcentrifuge tubes and immediately snap-freeze the vials in liquid nitrogen prior to re-storage at -70°C.

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service