Skip to Content
Merck
All Photos(2)

Key Documents

1.46787

Millipore

Tryptic Soy Agar - Ready-to-use Settle Plates

Lecithin, Tween®, Histidine, Sodium thiosulfate, ICR plus lockable plate, sterile; irradiated, plate diam. 90 mm, pkg of Triple packed, suitable for air monitoring

Synonym(s):

Soybean Casein Digest Agar, TSA,, Tryptic Soy Agar with LTHThio

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
CZK 2,070.00
1 EACH
CZK 12,400.00

CZK 2,070.00


Estimated to ship onMarch 12, 2025Details


Request more information

Select a Size

Change View
1 EACH
CZK 2,070.00
1 EACH
CZK 12,400.00

About This Item

UNSPSC Code:
41106217
NACRES:
NA.74

CZK 2,070.00


Estimated to ship onMarch 12, 2025Details


Request more information

material

polystyrene plate

Quality Level

Agency

EP
USP

sterility

sterile; irradiated

form

plate (ready to use culture media)

feature

ready-to-use

composition

Histidine
Lecithin
Sodium thiosulfate
Tween®

packaging

pkg of Triple packed

plate diam.

90 mm

application(s)

air monitoring
pharmaceutical

compatibility

for use with MAS-100®

storage temp.

2-25°C

suitability

nonselective for

Looking for similar products? Visit Product Comparison Guide

General description

Tryptic Soy Agar is a complex nutrient rich medium designed to isolate a wide range of bacteria, yeast and molds in clean rooms and isolators. The medium is supplemented with pyruvate, which efficiently neutralizes the presence of hydrogen peroxide used for decontamination of isolators. The neutralizers lecithin, polysorbate Tween®80, histidine, and sodium thiosulfate inactivate possible residues of active agents of disinfectants. In addition, the plate is supplied with a lockable lid (ICRplus;+) for safe transport after sampling without the risk of losing the lid. ICRplus plates are suitable for aerobic and anaerobic incubation by turning the lid into the close position for aerobic incubation and in the vent position for anaerobic incubation. The plates are triple bagged, and gamma irradiated in the final packaging and used for active and passive air monitoring, as well for personnel monitoring.

Application

Tryptic Soy Agar ICR+ settle plates are designed to determine the total aerobic and anaerobic microbial count in air (active and passive) inside isolators and clean rooms. The plates can also be used to monitor fingerprints of personnel.

Legal Information

MAS-100 is a registered trademark of MBV AG
TWEEN is a registered trademark of Croda International PLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Settle plates for isolators and critical cleanrooms

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service