Skip to Content
Merck

146753

IsoBag® Tryptic Soy Agar Contact Plus

190 mm alpha-port, 55 mm lockable Contact Plates, irradiated with Lecithin, Tween®, Histidine, and Sodium thiosulfate

Synonym(s):

TSA IsoBag®, Soybean Casein Digest Agar

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106231
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

EP, USP

sterility

sterile; γ-irradiated

form

plate (ready-to-use)

feature

2D bar code, ready-to-use

packaging

pkg of 100 plates

manufacturer/tradename

IsoBag®

D

55 mm

pH

7.3 ( in H2O)

application(s)

microbiology
pharmaceutical
surface monitoring

shipped in

ambient

storage temp.

15-25°C

suitability

bacteria, fungi

General description

The IsoBag® TSA Contact Plus is designed for quick and convenient transfer of culture media used for surface monitoring in isolators or RAB systems. The Beta bag consists of a sterile polyurethane DPTE Bag compatible with 190 mm Alpha Ports. The bag contains 100 lockable 55 mm contact plates filled with Tryptic Soy Agar.
TSA medium is used for the cultivation and isolation of a wide range of bacteria, yeasts, and molds. The formulation contains a combination of casein and soy peptones supplying organic nitrogen, particularly amino acids and longer-chained peptides. Sodium chloride maintains the osmotic equilibrium, and the medium is supplemented with neutralizers such as lecithin, polysorbate (Tween®) 80, histidine, and sodium thiosulfate to inactivate disinfectant residues. Additionally, the plate model (plus or “+”) is supplied with a lockable lid for safe transport after sampling.

Application

The IsoBag® TSA Contact plus is designed for quick and convenient transfer of culture media needed for surface monitoring in isolators or RAB systems.

Legal Information

ISOBAG is a registered trademark of Merck KGaA, Darmstadt, Germany
TWEEN is a registered trademark of Croda International PLC


Storage Class

11 - Combustible Solids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library