Sign In to View Organizational & Contract Pricing.
About This Item
NACRES:
NA.74
UNSPSC Code:
41106200
Bottle capacity:
250 mL
Bottle filling volume:
90 mL
Feature:
closure type blue screw cap with 3 loci, ready-to-use
Agency:
EP 2.6.1, JP 4.06, USP 71
Ph:
7.3
Quality Segment
agency
EP 2.6.1, JP 4.06, USP 71
sterility
sterile; autoclaved
form
liquid
shelf life
270 days
feature
closure type blue screw cap with 3 loci, ready-to-use
composition
Lecithin , Tween®
packaging
box of 6 bottles
bottle capacity
250 mL
bottle filling volume
90 mL
pH
7.3
application(s)
cosmetics
food and beverages
pharmaceutical
sterility testing
storage temp.
2-25°C
suitability
nonselective for
General description
Tryptic Soy Broth with neutralizers is a universal complex and highly nutritious medium and supports the growth of a wide variety of aerobic and facultative anaerobic microorganisms, including fungi. The neutralizing additives Polysorbate (Tween®) 80 and lecithin reverse the growth inhibitory effect of most disinfectants and antiseptics. Lecithin along with Polysorbate 80 is effective against quaternary ammonium compounds, amphoteric surfactants, benzamidines, chlorhexidines, and dequadin. Polysorbate 80 inactivates benzyl alcohol, carbanilides, dichlorbenzyl alcohols, benzoic acid, p-hydroxybenzoic acid and its esters, phenols, phenylethyl alcohols, and Solbrols (PHB esters).
Application
Tryptic Soy Broth with LT is used for the isolation and cultivation of fastidious aerobic bacteria, yeasts and molds in the presence of disinfectant residues. The medium designed for the testing of sterile and non-sterile substances, preparations, and products.
Features and Benefits
Our ready to use media provide the highest level of quality and testing confidence. They have been formulated and tested to meet the pharmacopoeia requirements. Sterility testing media and rinse solutions are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility and growth promotion tests. Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, therefore improving accuracy and significantly reducing the risk of incorrect interpretation and false results.
Legal Information
TWEEN is a registered trademark of Croda International PLC
Still not finding the right product?
Explore all of our products under Tryptic Soy Broth with Neutralizers
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Storage Class
12 - Non Combustible Liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.