Skip to Content
Merck

Skip To

1.46352

Yeast extract agar

ReadyTube®, bottle capacity 125 mL, bottle filling volume 100 mL, closure type, blue screw cap with 3 loci, box of 10 bottles

Synonym(s):

YE agar

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 ea
Please contact Customer Service for Availability
1 160,00 CZK

About This Item

NACRES:
NA.74
UNSPSC Code:
41106225

1 160,00 CZK


Please contact Customer Service for Availability

​
Request more information
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

ISO 6222

Quality Segment

form

solid

feature

closure type blue screw cap with 3 loci, ready-to-use

packaging

box of 10 bottles

manufacturer/tradename

ReadyTube®

bottle capacity

125 mL

bottle filling volume

100 mL

pH

7.2 ( in H2O)

application(s)

bioburden testing
food and beverages
industrial qc
water monitoring

storage temp.

2-25°C

suitability

nonselective for

General description

ReadyTube® is ready to use media to provide the highest level of quality and testing confidence for improving accuracy and significantly reducing the risk of false results. ReadyTube® Yeast extract agar is a highly nutritive medium recommended for the plate count of microorganisms in water. The media contains Yeast extract and peptone to provide nitrogenous compounds, vitamin B complex, and other growth nutrients. The Yeast extract agar formulation is in accordance to ISO 6222, the test strains according to ISO 11133 and released through an ISO 17025 accredited laboratory.

Application

ReadyTube® Yeast extract agar is a medium rich in nutrients, which permits the recovery of a wide spectrum of bacteria, yeast, and molds from water samples. It finds application in the food and beverage industry

Legal Information

READYTUBE is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Yeast extract agar


Storage Class

11 - Combustible Solids

WGK

nwg



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters