Skip to Content
Merck
All Photos(2)

Key Documents

1.46333

Millipore

Clear Thioglycollate Medium

bottle capacity 125 mL, bottle filling volume 100 mL, closure type, Red screw cap with septum and protector, pack of 10 bottles

Synonym(s):

Clear FTM, Clear Thioglycollate Broth, CTM, Fluid Thioglycollate Medium Clear

Sign Into View Organizational & Contract Pricing

Select a Size

10 X 100 ML
CZK 1,720.00

CZK 1,720.00


Estimated to ship onMarch 18, 2025Details



Select a Size

Change View
10 X 100 ML
CZK 1,720.00

About This Item

UNSPSC Code:
41106200
NACRES:
NA.74

CZK 1,720.00


Estimated to ship onMarch 18, 2025Details


Agency

EP 2.6.1
JP 4.06
USP 71

Quality Level

sterility

sterile; autoclaved

form

liquid (ready-to-use)

shelf life

12 mo.

feature

closure type Red screw cap with septum and protector
ready-to-use

packaging

pack of 10 bottles
single packed of (100 ml in 125 ml bottle with red screw cap and septum (10 bottles per box))

bottle capacity

125 mL

bottle filling volume

100 mL

pH

7.1

application(s)

cosmetics
pharmaceutical
sterility testing

storage temp.

2-25°C

suitability

aerobic bacteria
anaerobic bacteria

Looking for similar products? Visit Product Comparison Guide

General description

Fluid Thioglycollate Medium Clear is a universal complex medium. Thioglycollate and L-Cystine in the medium reduce the redox potential of the culture medium to create an anaerobic atmosphere. These agents also inactivate mercury and other heavy metal compounds. Resazurin indicates the reduction potential of the medium. Change of color from yellow to pink indicates an increase in oxygen concentration. Fluid Thioglycollate Medium Clear contains a reduced agar content, supplemented by a clear gelling agent, to offer the same media properties as FTM, with extra clarity.

Application

Fluid Thioglycollate Medium Clear is used for the isolation and cultivation of fastidious anaerobic as well as for aerobic microorganisms. This medium is suitable for sterility control of pharmaceutical products.

Features and Benefits

Our ready to use media provide the highest level of quality and testing confidence. They have been formulated and tested to meet the pharmacopoeia requirements. Sterility testing media and rinse solutions are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility and growth promotion tests. Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, therefore improving accuracy and significantly reducing the risk of incorrect interpretation and false results.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service