Skip to Content
Merck

Skip To

14-604-M

PI3 Kinase (p110δ/p85α) Protein, active, 10 µg

Active complex of N-terminal His6-tagged recombinant, full-length, human p110δ & untagged, recombinant, full length, human p85α. For use in Kinase Assays.

Synonym(s):

Active PI3 Kinase Protein, PI3 Kinase p110δ/p85α

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 μg

Available to ship TODAYfromAreál Kühne+Nagel spol. s r.o.

CZK 14,900.00
250 μg

Estimated to ship onJune 03, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 91,500.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32
eCl@ss:
32160405
Biological source:
human
Recombinant:
expressed in baculovirus infected Sf21 cells

CZK 14,900.00


Available to ship TODAYDetails


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

recombinant

expressed in baculovirus infected Sf21 cells

mol wt

Mw 121.6 kDa (P110δ), Mw 83.7 kDa (p85α)

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

General description

Complex of N-terminal His6-tagged recombinant, full-length, human p110δ and untagged, recombinant, full length, human p85α
Product Source: Coexpressed by baculovirus in Sf21 insect cells

Application

Active complex of N-terminal His6-tagged recombinant, full-length, human p110δ & untagged, recombinant, full length, human p85α. For use in Kinase Assays.
Research Category
Inflammation & Immunology

Biochem/physiol Actions

Protein Target: PI3Kδ/p85α
Target Sub-Family: PI3K/PI4K

Packaging

Also available in 250μg size (2x125μg)--call for pricing and availability and reference catalog number 14-604M when ordering the 250μg size.

Physical form

Ni2+/NTA-agarose

Preparation Note

6 months at -70°C

Analysis Note

routinely evaluated by the PIProfiler

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
14-603-M14-602-M14-790-M
biological source

human

biological source

human

biological source

human

biological source

-

technique(s)

activity assay: suitable (kinase)

technique(s)

activity assay: suitable (kinase)

technique(s)

activity assay: suitable (kinase)

technique(s)

activity assay: suitable (kinase)

recombinant

expressed in baculovirus infected Sf21 cells

recombinant

expressed in baculovirus infected Sf21 cells

recombinant

expressed in baculovirus infected Sf21 cells

recombinant

expressed in baculovirus infected Sf21 cells

UniProt accession no.

O00329

UniProt accession no.

P42338

UniProt accession no.

P42336

UniProt accession no.

P42336

mol wt

Mw 121.6 kDa (P110δ), Mw 83.7 kDa (p85α)

mol wt

Mw 124 kDa (p110β), Mw 83.7 kDa (p85&alpha)

mol wt

Mw 125.3 kDa (p110α), Mw 83.7 kDa (p85α)

mol wt

Mw 125.3 kDa (p110α), Mw 66.0 kDa (p65α)

NCBI accession no.

NM_005026.2

NCBI accession no.

NM_006219.1

NCBI accession no.

NM_006218.2

NCBI accession no.

NM_006218.2


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters