Skip to Content
Merck
All Photos(2)

Key Documents

1.46016

Millipore

Sabouraud Dextrose Agar

Tween®, Histidine, Sodium thiosulphate, Lecithin, lid, ICR: non lockable 30 mL, plate diam. 90 mm, sterile; γ-irradiated, triple packed of 20 or 120 plates, suitable for surface monitoring

Synonym(s):

ICR 30 mL plate, SDA, SDA with netralizers

Sign Into View Organizational & Contract Pricing

Select a Size

120 PKG
CZK 10,700.00

CZK 10,700.00


Estimated to ship onMarch 13, 2025Details



Select a Size

Change View
120 PKG
CZK 10,700.00

About This Item

UNSPSC Code:
41106200
NACRES:
NA.74

CZK 10,700.00


Estimated to ship onMarch 13, 2025Details


sterility

sterile; γ-irradiated

Quality Level

form

plate (ready-to-use)

feature

lid ICR: non lockable 30 mL
ready-to-use

composition

Histidine
Lecithin
Sodium thiosulphate
Tween®

packaging

triple packed of 20 or 120 plates

plate diam.

90 mm

pH

5.6 ( in H2O)

application(s)

microbiology
pharmaceutical

surface monitoring

compatibility

for use with Contact Plate

storage temp.

2-25°C

suitability

molds
yeasts

General description

Sabouraud Dextrose Agar + LTHTh - ICR in 90 mm settle plates (non-lockable) is a selective media for the determination of the total count of yeasts and molds in air via active or passive air monitoring as well as fingerprints of personnel in critical clean room classes or isolators. Sabouraud Dextrose Medium contains digests of animal tissues (peptones) which provide a nutritious source of amino acids and nitrogenous compounds for the growth of fungi and yeasts. Dextrose is added as the energy and carbon source. The low pH favours fungal growth. Lecithin, polysorbate (Tween®) 80, histidine and sodium thiosulfate are neutralizers to inactivate residues of disinfectants. In addition, the medium contains irradiation resistant antibiotics to inhibit the accompanying bacterial flora. To differentiate Tryptic Soy Agar (TSA) from Sabouraud Dextrose agar (SDA), SDA ICR media are filled in pink colored dish.

Application

Sab.Dex.A sel. +LTHTh - ICR is a selective medium for cultivation and isolation of yeasts and molds. The medium is used for environmental monitoring by determining the yeast and mold count in air through active or passive air monitoring as well as fingerprints of personnel in isolators and clean rooms.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service