Skip to Content
Merck
All Photos(1)

Key Documents

14-499-M

Sigma-Aldrich

Bmx Protein, active, 10 µg

Active, N-terminal His6 tagged, recombinant, full length, human Bmx, for use in Kinase Assays.

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
CZK 13,800.00
250 μG
CZK 89,400.00

CZK 13,800.00


Estimated to ship onApril 17, 2025Details


Request a Bulk Order

Select a Size

Change View
10 μG
CZK 13,800.00
250 μG
CZK 89,400.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

CZK 13,800.00


Estimated to ship onApril 17, 2025Details


Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in Sf21 cells

mol wt

Mw 79 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

Gene Information

human ... BMX(660)

General description

N-terminal His6 tagged, recombinant, full length, human Bmx

Biochem/physiol Actions

Protein Target: Bmx

Packaging

Also available in 250μg size as catalog # 14-499M - Call for pricing and availability.

Quality

routinely evaluated by phosphorylation of Poly (Glu4-Tyr) (4:1)

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service