Skip to Content
Merck
All Photos(1)

Key Documents

14-485-M

Sigma-Aldrich

IKKβ Protein, active, 10 µg

Active, recombinant, full-length human IKKβ, containing N-terminal His6 tag, for use in Kinase Assays.

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
CZK 13,900.00
250 μG
CZK 89,400.00

CZK 13,900.00


Estimated to ship onApril 08, 2025Details



Select a Size

Change View
10 μG
CZK 13,900.00
250 μG
CZK 89,400.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.26

CZK 13,900.00


Estimated to ship onApril 08, 2025Details


biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

mol wt

Mw 88 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

General description

Recombinant, full-length human IKKβ, containing N-terminal His6 tag

Biochem/physiol Actions

Protein Target: IKKβ

Packaging

Also available in 250μg size -- call for pricing and availability and reference catalog number 14-485M when ordering the 250μg size.

Quality

routinely evaluated by phosphorylation of IKKtide

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service