Skip to Content
Merck
All Photos(1)

Key Documents

14-347-M

Sigma-Aldrich

CHK2 Protein, active, 10 µg

Active, recombinant human CHK2, residues 5-543, containing N-terminal GST & C-terminal His-6 tags, for use in Kinase Assays.

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
CZK 13,800.00

CZK 13,800.00


Estimated to ship onMarch 24, 2025Details


Request a Bulk Order

Select a Size

Change View
10 μG
CZK 13,800.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

CZK 13,800.00


Estimated to ship onMarch 24, 2025Details


Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in E. coli

product line

Upstate®

specific activity

(For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.)

mol wt

Mw 89.6 kDa

purified by

(glutathione-agarose and Ni2+/NTA-agarose)

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

accession no.

 (FUNCTION: SwissProt: O96017 # Regulates cell cycle checkpoints and apoptosis in response to DNA damage, particularly to DNA double-strand breaks. Inhibits CDC25C phosphatase by phosphorylation on ′Ser-216′, preventing the entry into mitosis. May also play a role in meiosis. Regulates the TP53 tumor suppressor through phosphorylation at ′Thr-18′ and ′Ser-20′. )
NM_001005735.1

UniProt accession no.

Gene Information

human ... CHEK2(11200)

General description

Recombinant human CHK2, residues 5-543, containing N-terminal GST and Cterminal His6 tags

Biochem/physiol Actions

Protein Target: CHK2
Target Sub-Family: CAMK

Packaging

Also available in 10 μg size and in bulk
Also available in 250 µg size --call for pricing and availability and reference catalog number 14-347M when ordering the 250 µg size.

Quality

routinely evaluated by phosphorylation of the CHKtide substrate peptide

Target description

CHK2

Physical form

glutathione-agarose and Ni2+/NTA-agarose

Storage and Stability

1 year at -70°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Matsuoka et al.
Science (New York, N.Y.), 282(5395), 1893-1897 (1998-12-04)
In response to DNA damage and replication blocks, cells prevent cell cycle progression through the control of critical cell cycle regulators. We identified Chk2, the mammalian homolog of the Saccharomyces cerevisiae Rad53 and Schizosaccharomyces pombe Cds1 protein kinases required for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service