Skip to Content
Merck
All Photos(1)

Key Documents

14-253-M

Sigma-Aldrich

p38β2/SAPK2b2 Protein, active, 10 µg

Active, N-terminal GST fusion protein corresponding to full length human p38β2/SAPK2b2 activated with MKK6. For use in Kinase Assays.

Sign Into View Organizational & Contract Pricing

Select a Size

250 μG
CZK 89,400.00

CZK 89,400.00


Please contact Customer Service for Availability


Select a Size

Change View
250 μG
CZK 89,400.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

CZK 89,400.00


Please contact Customer Service for Availability

biological source

human

Quality Level

mol wt

Mw 71 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

General description

N-terminal GST fusion protein corresponding to full length human p38β2/SAPK2b2 activated with MKK6.
Product Source: full length human p38β2/SAPK2b2, expressed in E. coli.

Biochem/physiol Actions

Protein Target: p38β2/SAPK2b2
Target Sub-Family: CMGC

Quality

routinely evaluated by phosphorylation of myelin basic protein

Physical form

Affinity chromatography on glutathione-agarose beads

Storage and Stability

6 months at -20°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Cuenda et al.
The EMBO journal, 16(2), 295-305 (1997-01-15)
Stress-activated protein kinase-3 (SAPK3), a recently described MAP kinase family member with a wide-spread tissue distribution, was transfected into several mammalian cell lines and shown to be activated in response to cellular stresses, interleukin-1 (IL-1) and tumour necrosis factor (TNF)
A Cuenda et al.
FEBS letters, 364(2), 229-233 (1995-05-08)
A class of pyridinyl imidazoles inhibit the MAP kinase homologue, termed here reactivating kinase (RK) [Lee et al. (1994) Nature 372, 739-746]. We now show that one of these compounds (SB 203580) inhibits RK in vitro (IC50 = 0.6 microM)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service