1.41464
Parteck® MXP (Polyvinyl alcohol 4-88)
EMPROVE® ESSENTIAL, Ph. Eur., ChP, JPE, USP
Pharma Manufacturing
Synonym(s):
Parteck® Polyvinyl alcohol, PVA, PVOH
About This Item
Recommended Products
Agency
ChP
JPE
Ph. Eur.
USP
Quality Level
product line
EMPROVE® ESSENTIAL
form
fine powder
particle size
≤200 μm (d95)
mp
160-240 °C
application(s)
controlled release
pharmaceutical
solid formulation
solubility enhancement
storage temp.
15-25°C
InChI
1S/C2H4O/c1-2-3/h2-3H,1H2
InChI key
IMROMDMJAWUWLK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.
Application
Features and Benefits
- Superior supersaturation performance via precipitation inhibition
- High thermostability up to 250°C, extending the potential application range
- Stable high drug loads and suitability with a broad range of APIs
- Flexible release kinetics
- Compatible with several downstream processing capabilities, like direct compression, continuous manufacturing, thin film extrusion and 3D printing
- Optimized powder properties for excellent performance in the HME process
- Suitable polymer properties for HME, spray drying and 3D printing processes
Legal Information
Application
also commonly purchased with this product
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
3D printing can overcome challenges during formulation development, with a focus on enhancing API bioavailability in solid dispersions. Find out how in this technical article.
Our overview of API solubility and dissolution enhancement methods can help you decide between the many options to take during formulation.
Related Content
For selective laser sintering (SLS) polymers are needed for a successful workflow and selection of the best approach requires consideration of several factors.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service