Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
2.5 L | Estimated to ship TODAYfromAreál Kühne+Nagel spol. s r.o. | CZK 1,250.00 |
25 L | Estimated to ship TODAYfromAreál Kühne+Nagel spol. s r.o. | CZK 5,390.00 |
Estimated to ship TODAYfromAreál Kühne+Nagel spol. s r.o. | CZK 122,300.00 |
About This Item
Quality Segment
grade
GMO free, GMP, Halal, Kosher
agency
Ph. Helv.
vapor density
1.3 (vs air)
vapor pressure
3.23 psi ( 21.1 °C), 7.93 psi ( 37.7 °C)
product line
EMPROVE® EXPERT
bp
>100 °C (lit.)
density
1.18 g/mL at 20 °C
application(s)
cleaning in place
pharma/biopharma processes
foreign activity
Aflatoxin
storage temp.
2-25°C
SMILES string
Cl
InChI
1S/ClH/h1H
InChI key
VEXZGXHMUGYJMC-UHFFFAOYSA-N
General description
Packaging
1.37312.9025 / 25L / PE canister
1.37312.9208 / 200L / PE drum
1.37312.9950 / 950L / IBC
1.37312.9958 / 950L / IBC
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| application(s) cleaning in place | application(s) cleaning in place | application(s) cleaning in place | application(s) liquid formulation |
| foreign activity Aflatoxin | foreign activity Aflatoxin | foreign activity Aflatoxin | foreign activity - |
| agency Ph. Helv. | agency BP, ChP, JP, NF, Ph. Eur. | agency - | agency Ph. Helv. |
| product line EMPROVE® EXPERT | product line EMPROVE® EXPERT | product line EMPROVE® EXPERT | product line EMPROVE® ESSENTIAL |
| storage temp. 2-25°C | storage temp. 2-25°C | storage temp. 2-25°C | storage temp. 2-25°C |
| vapor density 1.3 (vs air) | vapor density 1.3 (vs air) | vapor density 1.3 (vs air) | vapor density 1.3 (vs air) |
Still not finding the right product?
Explore all of our products under Hydrochloric acid
signalword
Danger
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3
target_organs
Respiratory system
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
hcodes
Storage Class
8B - Non-combustible corrosive hazardous materials
Stay compliant with GMP using the Emprove® Program
Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.
Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.
Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite
Not finding what you are looking for? Visit the Emprove® Program Help page.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




